Order Entry
United States
ContactUsLinkComponent
Ammonia Buffer Solution pH10 (20 °C) for Complexometry, Fluka™, Honeywell Research Chemicals
Ammonia Buffer Solution pH10 (20 °C) for Complexometry, Fluka™, Honeywell Research Chemicals
Catalog # BJ33582-1L
CAS Number:  
Ammonia Buffer Solution pH10 (20 °C) for Complexometry, Fluka™, Honeywell Research Chemicals
Catalog # BJ33582-1L
Supplier Number:  33582-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Description:
    Ammonia buffer
  • Size:
    1 L
  • Cat. No.:
    BJ33582-1L
  • pH:
    10

Specifications

About this item