Order Entry
United States
Orders LinkContactUsLinkComponent
Ceramic Heat Pad

Ceramic Heat Pad

12" Square Ceramic Heat Pad
Catalog # 470313-334
Supplier:   GSC International
Supplier #:  CP-12
Ceramic Heat Pad
Catalog # 470313-334
Supplier:  GSC International
Ceramic Heat Pad
Ceramic Heat Pad

12" Square Ceramic Heat Pad

Price
$11.70
Log in for availability
Each Pack of 12
Product Details & Documents

These heat-insulating pads with low thermal conductivity are made from flexible aluminum and silica oxides with a small amount of organic binder.

  • Working temperature: 1000 °C or 1832 °F
  • Max. temperature: 1260 °C or 2300 °F
  • Sold as Each or as Pack of 12

They are useful for protecting surfaces when working with hot items like crucibles or flasks.

Specifications
  • Description:
    12" Square Ceramic Heat Pad