ContactUsLinkComponent
Dino-Lite Adjustable Precision Mount, Dunwell
  MSPP-RK-06A
Dino-Lite Adjustable Precision Mount, Dunwell
  MSPP-RK-06A
 :  RK-06A
  • Width:
    15 cm
  • Height:
    27 cm
  • Length:
    22 cm
  • Material:
    Metal
  • Cat. No.:
    MSPP-RK-06A
  • Weight:
    1060 g
The Dino-Lite tabletop stand with ESD protective base designed for general purpose, smooth focus adjustment and quick vertical movement.
  • Adaptable extension arms design add further mobility and flexibility to user’s applications
  • Compatible with all Dino-Lite handheld microscopes
  • Easy to assemble and disassemble
  • Highly flexible
Pairing the Dino-Lite with a stand minimizes interference with the microscope focus.
 : ROHS regulatory approval.
 : Includes stand with one year warranty period.