

Methanol, dry, Hydranal™ medium for volumetric one-component Karl Fischer titration, Fluka™
Supplier: Honeywell Research Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
:GHS08
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended after discussion
Specifications
Specification Test Results
- Suitability (for KF-titration):test passed
- Water (Karl Fischer):Max. 0.01 %
- Number Of Samples:Min. 2
Specification Test Results
Frequently Bought Together