Disodium hydrogen phosphate, anhydrous ≥98.0%, Reagent Grade
Catalog # 80731-078
Supplier: Avantor Performance Materials
CAS Number:
Specifications
- Size:25 kg
- Cat. No.:80731-078
- Supplier no.:VWRVMS0876-25KG
Specifications
Specification Test Results
- Assay:Min. 98.0%
- Appearance:White Crystalline Powder
- Solubility in water (1 g/10 ml):Clear and colorless
- pH (1%):8.7 - 9.6
- Heavy metals (as Pb):Max. 10 ppm
- Insoluble Matter:Max. 0.2%
- Loss on Drying:Max. 5%
Specification Test Results
About this item
:
:141.96 g/mol
Storage Temperature:Ambient
CAS Number:7558-79-4
Formula:Na₂HPO₄
MDL Number:MFCD00003496