Urea Agar Base (Christensen), HiMedia Laboratories
Supplier: HiMedia
Certificates
About this item
Urea Agar Base is recommended for the detection of urease production, particularly by Proteus vulgaris, Micrococci and
Paracolon organisms.
- Light pink colored homogeneous free flowing powder
- Firm gelling, comparable with 1.5% agar gel
- Cultural characteristics can be observed after an incubation at 35…37 °C for 18 to 24 hours
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.