Phosphate Buffer Solution pH 7,00 (20 °C), Fluka™, Honeywell Research Chemicals
BJ73173-1L
:
- Pack type:Plastic bottle
- Description:Phosphate buffer solution
- Size:1 L
- Cat. No.:BJ73173-1L
- pH:7.00
- pH accuracy:±0.02
Potassium dihydrogen phosphate / di-sodium hydrogen phosphate