HiLoad 16/600 Superose 6 pg Preparative SEC Column, Cytiva
Supplier: Cytiva
Certificates
About this item
HiLoad 16/600 Superose 6 prep grade (pg) is a prepacked size exclusion chromatography (SEC) column for high-resolution purification of large biomolecules and protein complexes. The column is an excellent choice for purification of sample volumes up to 5 ml.
- Convenience and reproducibility: can be used with standalone pump or a chromatography system
- High-resolution separations: purification is possible across an exceptionally broad molecular-weight range
- Scalable: separations can be scaled up using Superose 6 prep grade resin
- ÄKTA system compatible: fittings (¹/₁₆") for direct connection to ÄKTA systems and other chromatography systems
This column is packed with Superose 6 pg resin, which provides good resolution separations of proteins and other biomolecules according to size. The wide fractionation range of the resin enables reproducible and robust separations in the molecular weight range from 5000 up to 5000000.
Specifications
- Catalog No:
- 76308-980
- Storage:
- 4...30 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Frequently Bought Together