Specifications
- Description:Low Toxic Silicone Adhesive, 5 mL Syringe
- Cat. No.:MSPP-KWIK-SIL
Specifications
About this item
Bio-compatible adhesive for live tissue and nerve studies. Ideal for neuroscience applications, nerve studies and more.
- Pre-mixing tips simplify use (Includes 10 mixing tips)
- Medium strength adhesion
- Rapid curing silicone adhesive, cure on contact
- Cures without producing heat
- Kwik-Sil dries clear;
- Kwik-Sil dries a little faster than Kwik-Cast
Kwik-Sil™ is a translucent, medium-viscosity silicone adhesive, developed for chronic peripheral nerve studies such as anterograde tracing with fluorescent indicators or electrode recording. Good adhesion and mechanical properties (tear strength and elongation) allow days of study without breaking of the bonding. Curing speed is very reproducible.
Kwik-Sil silicones have very low toxicity before, during and after curing. The by-product of curing is a small amount of hydrogen gas, which is much less toxic to cells than acetic acid or alcohol from traditional RTV silicone systems.
Kwik-Sil curing speed is hundreds of times faster than traditional RTV silicones. A curing time of a few minutes at room temperature is especially useful for encapsulation of live tissue or implanting into a live animal. Unlike many vinyl-based silicones in which the platinum complex catalysts are easily poisoned by contamination from amines and animal tissue, Kwik-Sil and Kwik-Cast are not sensitive to contamination from animal tissue.