Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Temperature range:40...65 °C (paraffin)
- Description:Leica ASP300 S fully enclosed tissue processor, 4.3 L (reagent bottle volume)
- Cat. No.:76001-002
- W×D×H:59×68×132 cm (24×27×52")
- Weight:160 kg (352.74 lbs.), instrument only
Specifications
About this item
The Leica ASP300 S is a freestanding, fully- enclosed vacuum tissue processor for programmable processing of histological tissue specimens in the fields of pathology and clinical histology.
- RemoteCare - optional remote diagnostic via Internet interface
- User access control and monitoring via multiple-step password protection
- Large, high-quality color LCD touchscreen, solvent-resistant
- Intelligent user interface (protecting against programming and operation errors)
- Intelligent Reagent Management System - RMS (can be switched ON/OFF)
- Export function of RMS data to standard spreadsheet programs (Excel or others)
- Labels for reagent bottles with 'reagent group' color coding for different reagent types and concentrations
- Microwave-resistant plastic baskets for cassettes
- Powerful stirrer unit for efficient infiltration and even reagent temperatures throughout the retort
- 'Fast' infiltration protocols for urgent biopsies
- Funnel for splash-free filling of the reagent bottles
The instrument provides three cleaning stations plus an optional connection for an external cleaning station. The 4.5-liter processing retort is made of electropolished stainless steel for easy cleaning with a contact-free optical sensor to check the fluid level. Retort capacity for up to 300 standard cassettes when using metal baskets or 252 standard cassettes when using the microwave resistant basket set for ASP300 S (optional accessory).
A selection of four further cleaning programs, which can be customized by the operator, are available. An easy-to-use remote reagent handling system can drain and refill reagent bottles from bulk containers via the retort and a hoseplugged into the processor module. The 3-step retort draining system is tailored to minimize reagent carry-over.
For options, contact your VWR representative.