C-Flex® Animal-Derived Component Free Tubing, Cole Palmer
Supplier: Avantor
Certificates
About this item
C-Flex® is a clear, transparent tubing, suitable for critical applications.
- Tensile and tear strength
- Biocompatibility
- Chemical resistance
- Low gas permeability
- Smooth surface
These tubes can be sterilized by autoclave, ethylene oxide or gamma irradiation.
The temperature range is –45 to +135 °C.
Certifications: Resin meets USP and Class VI requirements. Tubing is manufactured according to GMP.
Frequently Bought Together