Order Entry
United States
ContactUsLinkComponent
Pluronic® F-127 20% in DMSO
Pluronic® F-127 20% in DMSO
Catalog # 89139-288
Supplier:  Biotium
CAS Number:  
Pluronic® F-127 20% in DMSO
Catalog # 89139-288
Supplier:  Biotium
Supplier Number:  59004
CAS Number:  

Specifications

  • Size:
    1 ml
  • Cat. No.:
    89139-288
  • Supplier no.:
    59004

Specifications

About this item

 :
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Number:MFCD00082049