ContactUsLinkComponent
Warmbier SRM110 Pocket Digital Surface Resistance Meter, Transforming Technologies
  76210-060
undefined
Warmbier SRM110 Pocket Digital Surface Resistance Meter, Transforming Technologies
  76210-060
 :  7100.SRM110
  • Description:
    Warmbier SRM110 Pocket Digital Surface Resistance Meter
  • Cat. No.:
    76210-060
Quickly and accurately take resistance measurements with the Warmbier SRM110.
  • LED readout will report the resistance in powers of 10
  • Indicates if the surface is insulative, conductive, or dissipative
  • Test range: 10³ to 10¹²
  • Test voltage: 10 V/100 V (automatic ranging)
  • Accuracy + 5%
  • Built-in resistivity probes
The Warmbier SRM110 meter uses parallel electrodes and concentric rings to accurately measure RTT, RTG, or resistivity for periodic verification, factory audits, or test lab product evaluations. This meter is designed to verify incoming materials such as floor mats, table mats, bins, and shelving in static controlled areas. Work surfaces, bags, wrist straps, footwear, packaging materials, garments, and ESD chairs can also be tested with the device.
 : SRM110 meets periodic test requirements per Compliance Verification ESD TR53 and conforms to ANSI/EOS/ESD (S4.1, S7.1, S12.1, S2.1).
 : Supplied with calibration certificate in a carrying case.