Order Entry
United States
ContactUsLinkComponent
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 89139-274
Supplier:  Biotium
CAS Number:  
undefined
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥95% (by HPLC) fluorescent indicator, for chloride determination
Catalog # 89139-274
Supplier:  Biotium
Supplier Number:  52011
CAS Number:  

Specifications

  • Size:
    100 mg
  • Cat. No.:
    89139-274
  • Supplier no.:
    52011

Specifications

About this item

 :
 :326.19 g/mol
 :GHS07
CAS Number:162558-52-3
Formula:C₁₄H₁₆NO₃Br
MDL Number:MFCD00467854