Order Entry
United States
ContactUsLinkComponent
PMA Enhancer for Gram Negative Bacteria solution 5X, Biotium
PMA Enhancer for Gram Negative Bacteria solution 5X, Biotium
Catalog # 10154-206
Supplier:  Biotium
CAS Number:  
PMA Enhancer for Gram Negative Bacteria solution 5X, Biotium
Catalog # 10154-206
Supplier:  Biotium
Supplier Number:  31038
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    PMA Enhancer for Gram Negative Bacteria solution 5X
  • Size:
    16 ml
  • Cat. No.:
    10154-206
  • Supplier no.:
    31038

Specifications

About this item

PMA Enhancer for Gram Negative Bacteria was designed to improve PMA- and PMAxx™-mediated discrimination between live and dead gram-negative bacteria.

  • Clear liquid
  • Should be stored at 4°C. When stored as recommended, the product is stable for at least six months from date of receipt.

PMA is a high affinity photoreactive DNA binding dye developed by Biotium for viability PCR (40019). PMAxx™ is our new and improved viability PCR dye (40069). The dyes are weakly fluorescent by themselves but become highly fluorescent upon binding to nucleic acids. Upon photolysis, the dye forms a stable covalent bond with the DNA, thus resulting in permanent DNA modification. The dye is cell membrane-impermeable and thus can be used to selectively modify DNA from dead cells with compromised membrane integrity, while leaving DNA from viable cells intact. PMA and PMAxx™ inhibit PCR amplification of modified DNA templates. Consequently the dye is useful in the selective detection of viable pathogenic cells by quantitative real-time PCR.

PMA Enhancer for Gram Negative Bacteria was designed to improve PMA- and PMAxx™-mediated discrimination between live and dead gram-negative bacteria. PMA Enhancer is provided as a 5X solution, and is added to a sample before the addition of dye. When a sequence from a gram-negative bacteria is amplified by PCR, samples pre-treated with Enhancer show a decrease in the signal from dead cells, with no change in the signal from live cells. Thus, PMA or PMAxx™ plus Enhancer is the optimal way to perform viability PCR on gram-negative bacteria.
The mechanism by which Enhancer improves live/dead discrimination has not been determined. It may improve passive permeability of dead cell walls or membranes to the dye, and/or improve access of the dye to the dead cell DNA.

Note: PMA Enhancer is not intended for use when gram-positive bacteria are to be detected. PMA Enhancer may adversely affect the amplification of live cell DNA from gram-positive bacteria.

For photoactivation of PMA, PMAxx™ or EMA dye, we recommend the use of Biotium’s PMA-Lite™ LED Photolysis Device (E90002), which is designed to conduct photolysis under controlled conditions.