Order Entry
United States
ContactUsLinkComponent
Tetrahydrofuran ≥99.8% (by GC-FID), universal grade for HPLC, for gas chromatography, for UV spectrophotometry
Tetrahydrofuran ≥99.8% (by GC-FID), universal grade for HPLC, for gas chromatography, for UV spectrophotometry
Catalog # 85003-814
Supplier:  BDH
CAS Number:  
undefined
Tetrahydrofuran ≥99.8% (by GC-FID), universal grade for HPLC, for gas chromatography, for UV spectrophotometry
Catalog # 85003-814
Supplier:  BDH
Supplier Number:  BDH6040-4
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    Case of 4
  • Environmentally Preferable:
  • Cat. No.:
    85003-814

Specifications

Specification Test Results

  • Assay (by GC-FID):
    Min 99.8 %
  • Color:
    Max 10 APHA
  • Peroxides/Epoxides (as Hydrogen peroxide at time of packaging):
    Max 0.015 %
  • Residue after evaporation:
    Max 5 ppm
  • Titrable acid:
    Max 0.0008 meq/g
  • Water (by KF coulometry):
    Max 0.03 %
  • 212 nm:
    Max 1.00 AU
  • 225 nm:
    Max 0.50 AU
  • 250 nm:
    Max 0.17 AU
  • 350 nm:
    Max 0.01 AU

Specification Test Results

About this item

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide
 :
 :72.11 g/mol
EINECS:203-726-8
UN:2056
REACH:05-2114615188-48
 :GHS02
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:109-99-9
Formula:C₄H₈O
MDL Number:MFCD00005356
Boiling Pt:66
Melting Pt:-108.5
Density:0.888 g/cm³ (20 °C)
Flash Pt:–21.5 °C
ADR:3 II
Merck Index:12,09351