Dissipative ESD Trash Can Liners, Transforming Technologies
Safely dispose of waste materials that pose a static damage threat.
- Dissipative trash liners for ESD protection
Translucent pink Anti-Static Trash Can Liners are ideal for use in cleanrooms, manufacturing, inspection, quality control, and research industries where static control is crucial. The average surface resistivity of the liners less than or equal to 10¹¹ ohms per square, with a static decay rate less than two seconds.
: Meets or exceeds requirements of ANSI ESD-S20.20
Frequently Bought Together