ESD Safe Tray Liners, Transforming Technologies
About this item
ESD Safe Tray Liners are designed for use in lining the surface of process trays to keep items protected and stable.
- ESD dual-layer rubber construction
- Dissipative top layer and a conductive bottom layer
- Multiple color options available
ESD rubber provides a better path-to-ground and cushion versus the hard surface of the conductive tray. The economical dual-layer rubber material is cut to fit a 16×24" process tray. Each 0.080" thick ESD tray liner has a dissipative top layer and a conductive bottom layer.
Certifications: Liners meet or exceeds requirements of ANSI ESD S541-2003. RoHS compliant
4 Options Available Below
Product Details & Documents
ESD Safe Tray Liners are designed for use in lining the surface of process trays to keep items protected and stable.
- ESD dual-layer rubber construction
- Dissipative top layer and a conductive bottom layer
- Multiple color options available
ESD rubber provides a better path-to-ground and cushion versus the hard surface of the conductive tray. The economical dual-layer rubber material is cut to fit a 16×24" process tray. Each 0.080" thick ESD tray liner has a dissipative top layer and a conductive bottom layer.
Certifications: Liners meet or exceeds requirements of ANSI ESD S541-2003. RoHS compliant
Product Family Options
Product Information