ESD Safe SMD Component Storage Box Cabinet, Transforming Technologies
Catalog # 76210-002
Supplier: Transforming Technologies
Specifications
- Description:6-Drawer ESD Safe SMD Component Storage Box Cabinet, black
- Cat. No.:76210-002
- Ext. dimensions:265×355×205 mm
Specifications
About this item
Organize, protect, and store interlocking SMD boxes in this six-drawer conductive cabinet.
- Made of volume loaded conductive polystyrene
- Tray compatible size combination
- Spring loaded lid
- Recyclable
The six-drawer cabinet allows for common storage of ESD safe boxes. Specifically compatible with the SM0880 and SM0870 series SMD boxes.
Certifications: Complies with EN 61340-5-1. Meets or exceeds requirements of ANSI ESD-S20.20