ContactUsLinkComponent
Ptec™ AirFLEX Flexible Neck Ionizing Nozzle, Transforming Technologies
Ptec™ AirFLEX Flexible Neck Ionizing Nozzle, Transforming Technologies
Catalog # 76209-930
undefined
Ptec™ AirFLEX Flexible Neck Ionizing Nozzle, Transforming Technologies
Catalog # 76209-930
Supplier Number:  IN4200
Specifications
  • Description:
    Ptec™ AirFLEX flexible neck ionizing nozzle
  • Environmentally Preferable:
  • Cat. No.:
    76209-930
  • Electrical:
    100/240 V, 50/60 Hz
Specifications
About this item
The IN4200 flexible nozzle neck frees the operator’s hands during assembly and manufacturing processes. A foot pedal controls both ionization and airflow, which reduces compressed air costs and extends the life of the ionizer.
  • Flexible neck allows user to optimally position airflow
  • Multiple nozzle options to customize airflow for specific applications
  • Hands-free operation with foot pedal control
  • Ionization (green) and HV alarm (red) lights
  • No calibration required
The model IN4000 ionizing air nozzle produces a 68 KHz AC output of approximately 2200 V and a continuous stream of balanced air ions. AirFLEX ionizers are valuable because they can be mounted in hard to reach areas that larger ionizers just cannot handle. The IN4200 is designed to neutralize electrostatic charges in benchtop environments, sensitive materials assembly, medical device, SMT, electronics assembly, automotive packaging, cleanroom, and laboratory environments.