Specifications
- Description:StaticAIRE™ still air ionizer
- Cat. No.:76209-984
- Electrical:110 - 240 V, 50/60 Hz
Specifications
About this item
The StaticAIRE™ Still Air Ionizer quickly and effectively eliminates static without using airflow. It’s designed for use in a variety of localized applications where space is limited and is ideal for installation inside equipment.
- Quickly eliminate static charges in localized areas without airflow
- Effective ionization range: 12"
- Ideal where space is limited and installed inside equipment
- Easy adjustment and calibration
An ideal product for mini-environments, analytical laboratories, and cleanrooms, the StaticAIRE™ Ionizer was designed to be virtually maintenance-free. It is factory set to achieve a maximum balanced ion output in standard, non-air assisted applications. Balance and pulse rate can be adjusted quickly to achieve optimal performance with trimpots located on the front of the ionizer. The IN1000 ionizer comes factory calibrated and with a calibration certificate. Ionizer performance can be tested and verified with a charge plate monitor.