Disposable ESD Heel Grounder, Transforming Technologies
Catalog # 76209-916
Supplier: Transforming Technologies
Specifications
- Color:Yellow
- Description:Disposable ESD Heel Grounder
- Cat. No.:76209-916
Specifications
About this item
Disposable Heel Grounder is the perfect solution for plant visitors and other temporary personnel who enter static controlled environments with ESD protective flooring.
- Low cost method of grounding temporary personnel
- High visibility yellow
- Disposable design is ideal for one-time use
Constructed of a durable non-woven fabric impregnated with carbon, disposable heel grounders are comfortable to wear, easily identified, and use the entire volume of the fabric to provide protection.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20