Low Profile Common Point Ground Cords, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Low profile common point ground cords connect multiple aspects of an ESD workstation to ground with a smaller connection point.
- Common point ground cord for two wrist straps and ESD mat
- One meg resistor for safety
Highly durable and reliable Low Profile Common Point Ground (CPG) Cords are equipped with two standard banana jacks, and will ground up to two operators at the same time. The low profile common point ground includes a 15’ long black PVC cord made with 7 strand tinsel wire to #10 ring terminal. The molded connection point houses a 1 megOhm 1/4 watt resistor and attaches to table mats via a 10 mm stud.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Frequently Bought Together