Anti-Static Cellulose Tapes, Transforming Technologies
Supplier: Transforming Technologies
Certificates
About this item
Available with or without a printed ESD symbol, anti-static clear cellulose tape is a general-purpose high tack tape with excellent holding power.
- Charge decay of <0.014 seconds
- Solvent based natural rubber adhesive
- 35 µm pure cellulose backing
Ideal for general packaging in environments where static is an issue, this tape can be used to seal packages, containers, or supplies in manufacturing. Perfect for the electronics industry because the pressure sensitive tape is virtually static-free and can be used to close inner boxes and wrap electronic components. It is hand tear-able and biodegradable.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Frequently Bought Together