ContactUsLinkComponent
QC Colloidal Coomassie Stain, Bio-Rad
Catalog # 102827-652
Supplier:  Bio-Rad
CAS Number:  
QC Colloidal Coomassie Stain, Bio-Rad
Catalog # 102827-652
Supplier:  Bio-Rad
Supplier Number:  1610803
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    QC Colloidal Coomassie Stain
  • Size:
    1 L
  • Cat. No.:
    102827-652
  • Supplier no.:
    1610803
Specifications
About this item
Ready-to-use, non-hazardous colloidal Coomassie G-250 stain for protein polyacrylamide gels. Use QC Colloidal Coomassie to stain proteins in SDS-PAGE gels without using methanol for fixing.
  • Low background, high sensitivity, superior reproducibility
  • Environmentally friendly formulation — no addition of methanol or acetic acid required; eliminates the need for hazardous waste disposal
  • Flexible staining and destaining times — from 1 hour to overnight
  • No alcohol addition or dilution steps when staining polyacrylamide gels
  • One-part, ready-to-use colloidal Coomassie stain