ContactUsLinkComponent
Ward's® Properties of Aspirin (Aspirin Free)
Ward's® Properties of Aspirin (Aspirin Free)
Catalog # 470313-054
Supplier:  Ward's Science
undefined
Ward's® Properties of Aspirin (Aspirin Free)
Catalog # 470313-054
Supplier:  Ward's Science
Supplier Number:  AD-KK9015-KIT
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Properties of Aspirin (Aspirin Free)
  • No. of lab groups:
    15
  • Cat. No.:
    470313-054
Specifications
About this item
What is the difference between Aspirin pills?
  • Kit includes all necessary materials to perform activity except aspirin tablets
  • Student study guide copymasters and teacher manual included
Aspirin is to this day the most widely used painkiller and largest selling non-perscription medicine in the world. Learn about the history of the development of acetylsalicylic acid (aspirin) and test the performance of several different types of aspirin. Students will examine the solubility of aspirin in varying gastrointestinal environments, a comparison of active ingredients in regular and extra-strength aspirin, and the differences between regular and buffered aspirin.
Certifications: Aligned to the Next Generation Science Standards (NGSS): PS1.A, PS1.B; MS-PS1-2; MS-PS1-3.
Caution: This product can expose you to chemicals including Phenolphthalein/Methanol/Methyl Isobutyl ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.