Order Entry
United States
ContactUsLinkComponent
Propionic acid ≥99% (by GC), J.T.Baker®
Propionic acid ≥99% (by GC), J.T.Baker®
Catalog # JTU340-09
CAS Number:  
MOC:  RegulatedCertificates
undefined
Propionic acid ≥99% (by GC), J.T.Baker®
Catalog # JTU340-09
Supplier Number:  U340-09
CAS Number:  
MOC:  RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    430 lbs.
  • Cat. No.:
    JTU340-09

Specifications

Specification Test Results

  • Assay (by GC) (CH₃CH₂COOH):
    ≥99 %

Specification Test Results

About this item

 :
 :74.08 g/mol
Merck Index:13,07917
Boiling Pt:141 °C (1013 hPa)
Melting Pt:–21 °C
Density:0.99 g/cm³ (20 °C)
Flash Pt:49 °C
Storage Temperature:Ambient
EINECS:201-176-3
UN:3463
ADR:8 (3),II
 :Management of Change (MOC) category = R
REACH:01-2119486971-24
 :GHS02
 :GHS05
 :GHS06
CAS Number:79-09-4
Formula:C₃H₆O₂
MDL Number:MFCD00002756