5-[(4-Propyloxyphenyl)methylene]-2,4-thiazolidinedione (SMI-16a) ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
CAS Number:587852-28-6
:
Formula:C₁₃H₁₃NO₃S
MDL Number:MFCD31382191
:263.31 g/mol
Storage Temperature:Freezer
Frequently Bought Together