ContactUsLinkComponent
Leybold Determination of the Molar Mass of Gases
Leybold Determination of the Molar Mass of Gases
Catalog # 470311-216
Supplier:  Klinger
undefined
Leybold Determination of the Molar Mass of Gases
Catalog # 470311-216
Supplier:  Klinger
Supplier Number:  C1.1.1.2
Specifications
  • Description:
    Determination of the Molar Mass of Gases
  • Cat. No.:
    470311-216
Specifications
About this item
Leybold Chemistry Experiments.
  • Show that gases also have a weight
  • Determine the molecular mass of gases
  • Learn about the molar volume
At constant pressure and constant temperature, any gas occupies the same volume regardless of the type of atom or the composition. So if we know the volume, pressure and temperature, we can then determine the molar mass of gases. The measurement is conducted in experiment C1.1.1.2 with a glass sphere for weighing gases.