Recording of Absorption Spectra, Leybold Products
470311-210
: Klinger
- Description:Recording of Absorption Spectra
- Cat. No.:470311-210
Leybold Chemistry Experiments.
- Observe and understand light spectra
- Recognize connection between absorption, reflection and transmission
- Appreciate color as light of characteristic wavelengths
- Identify the interaction of light with colored liquids
In experiment C3.3.1.2, the solution of a dye is exposed to the light from a lamp. For the absorption spectrum, the light passing through the colored solution is recorded with a spectrometer. The absorption spectrum is compared with the continuous spectrum of the light from the lamp.