ContactUsLinkComponent
Thermal Analysis of Hydrocarbons, Leybold Products
Thermal Analysis of Hydrocarbons, Leybold Products
Catalog # 470311-206
Supplier:  Klinger
undefined
Thermal Analysis of Hydrocarbons, Leybold Products
Catalog # 470311-206
Supplier:  Klinger
Supplier Number:  C2.1.2.1
Specifications
  • Description:
    Thermal Analysis of Hydrocarbons
  • Cat. No.:
    470311-206
Specifications
About this item
Leybold Chemistry Experiments.
  • Composition of organic compounds
  • Reactions in organic chemistry
  • Petrochemistry
  • Synthesis and purification of organic compounds
In order to determine the number of hydrogen atoms as compared with the number of carbon atoms, hydrocarbons can be separated into their elements by thermal decomposition. With gaseous hydrocarbons, this can be done with a filament in a combustion chamber. There the gas is split into hydrogen and carbon black (soot). The ratio of hydrogen to carbon in the compound can be derived directly from the increase in gas volume. In experiment C2.1.2.1, the gases methane and ethane are studied in this way.