Order Entry
United States
ContactUsLinkComponent
Crystal violet 1% in aqueous solution stain for bacteriology (Brown-Hopps method)
Irritant
Health
Crystal violet 1% in aqueous solution stain for bacteriology (Brown-Hopps method)
Catalog # 100504-496
Minority or Woman-Owned Business Enterprise
CAS Number:  
Irritant
Health
Crystal violet 1% in aqueous solution stain for bacteriology (Brown-Hopps method)
Catalog # 100504-496
Supplier Number:  26106-01
Minority or Woman-Owned Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 ml
  • Cat. No.:
    100504-496
  • Supplier no.:
    26106-01

Specifications

About this item

 :
 :407.99 g/mol
 :GHS07
 :GHS08
CAS Number:548-62-9
Formula:C₂₅H₃₀ClN₃
MDL Number:MFCD00011750