J.T.Baker®, Silica Gels for Preparative Chromatography, Low Pressure
These are used in low pressure preparative chromatography. The mesh specifications ensure there is no excess of fines in the product which can affect flow rates and separation performance.
- High surface area improves productivity
- Narrow pore size distribution sharpens peaks and improves resolution
Frequently Bought Together