Basic fuchsin in alcohol reagent A for PREVI® COLOR GRAM instrument
Catalog # 89127-612
Supplier: bioMerieux
CAS Number:
Specifications
- Size:500 ml
- Cat. No.:89127-612
- Supplier no.:29585
Specifications
About this item
:
:337.85 g/mol
:GHS02
:GHS07
CAS Number:632-99-5
:58969-01-0
Formula:C₂₀H₂₀ClN₃
MDL Number:MFCD00012569