Setup/Stab/Masscal Solution for ELAN 9000/6100, PerkinElmer
Catalog # 97011-972
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:10 μg/L (Ba, Cd, Ce, Cu, In, Pb, Mg, Rh, U)
- Matrix:1% HNO₃
- Description:Setup/Stab/Masscal Solution
- Size:1000 ml
- Environmentally Preferable:
- Cat. No.:97011-972
- Supplier no.:N8125030
Specifications
About this item
Inorganic solution is for use with the ELAN 9000/6100 in atomic spectroscopy.