Order Entry
United States
ContactUsLinkComponent
Antifoam C Medical Emulsion, Biotech Reagent, J.T.Baker®
Antifoam C Medical Emulsion, Biotech Reagent, J.T.Baker®
Catalog # JT8535-02
Supplier:  Avantor
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates
undefined
Antifoam C Medical Emulsion, Biotech Reagent, J.T.Baker®
Catalog # JT8535-02
Supplier:  Avantor
CAS Number:  
MOC:  Non-Regulated, With InformationCertificates

Specifications

  • Size:
    1 L
  • Cat. No.:
    JT8535-02
  • Supplier no.:
    8535-02

Specifications

Specification Test Results

  • Assay:
    25.5 - 33.0 %
  • Appearance:
    Passes Test
  • Microbial Limit (per g):
    ≤ 100
  • Defoaming Activity (seconds):
    ≤ 15
  • Heavy Metals(as Pb) (/ug/g):
    ≤ 5
  • Identification (by IR):
    Passes Test

Specification Test Results

About this item

 :
 :Management of Change (MOC) category = NI