Order Entry
United States
ContactUsLinkComponent
Minum-Ware® Air Reflux Condenser, Chemglass
Minum-Ware® Air Reflux Condenser, Chemglass
Catalog # 80064-116
Supplier:  Chemglass
Small Business Enterprise
undefined
Minum-Ware® Air Reflux Condenser, Chemglass
Catalog # 80064-116
Supplier:  Chemglass
Supplier Number:  MW-40-01
Small Business Enterprise

Specifications

  • [ST] joint:
    14/10
  • Height:
    145 mm
  • Length:
    100 mm
  • Cat. No.:
    80064-116
  • Supplier no.:
    MW-40-01

Specifications

About this item

Condenser uses the surrounding air to dissipate the heat of the reaction during refluxing.

  • Features a top threaded outer joint and lower inner drip joint
  • All joints are 14/10
Delivery: Supplied with open-top screw cap and Viton O-ring.