Determining Wavelengths from Balmer, Leybold Products
470311-142
: Klinger
- Description:Determining Wavelengths from Balmer
- Cat. No.:470311-142
Leybold chemistry experiments.
- X-ray physics
- Radioactivity
- Nuclear physics
- Quantum physics
In the experiment P6.2.1.1, the emission spectrum is excited using a Balmer lamp filled with water vapor, in which an electric discharge splits the water molecules into an excited hydrogen atom and a hydroxyl group. The wavelengths of the lines Hα, Hb and Hγ are determined using a high-resolution grating.