Determining Planck's Constant, Leybold Products
Catalog # 470311-138
Supplier: Klinger
Specifications
- Description:Determining Planck's Constant
- Cat. No.:470311-138
Specifications
About this item
Leybold chemistry experiments.
- Millikan experiment
- Dual nature of wave and particle
- Atomic shell
- X-ray physics
In the experiment P6.1.4.3, the capacitor method described previously (see P6.1.4.1) is used to generate the opposing voltage U between the cathode and the anode of the photocell. The voltage at the capacitor is measured without current using the electrometer amplifier.