Order Entry
United States
ContactUsLinkComponent
Human Recombinant IL-28A/IFN-lambda 2 (from Embryonic kidney cell)
Human Recombinant IL-28A/IFN-lambda 2 (from Embryonic kidney cell)
Catalog # 103705-716
Supplier:  Novus Biologicals
CAS Number:  
Human Recombinant IL-28A/IFN-lambda 2 (from Embryonic kidney cell)
Catalog # 103705-716
Supplier:  Novus Biologicals
Supplier Number:  8417-IL-025/CF
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    Embryonic kidney cell
  • Species:
    Human
  • Size:
    25 µg
  • Endotoxin Content:
    <0.10 EU/μg of protein
  • Form:
    Lyophilized
  • Gene ID:
    IFNL2
  • Carrier-Free:
    Y
  • Animal-Free:
    Y
  • Protein Synonyms:
    lambda 2|IL28A|Interleukin-28A|Interferon|IFN-lambda 2
  • Accession No.:
    Q8IZJ0
  • Protein/Peptide Name:
    IL-28A/IFN-lambda 2
  • Purity:
    >95%
  • Molecular Weight:
    20 - 23 kDa
  • Tested Applications:
    Bioassay
  • Cat. No.:
    103705-716

Specifications

About this item

IL‑28A (Interferon-lambda 2; IFN-lambda 2), IL-28B/IFN-lambda 3, and IL-29/IFN-lambda 1 are type III interferons which are distantly related to IL-10 family and type I IFN family cytokines. Mature human IL-28A is an approximately 22 to 25 kDa protein that shares 66% amino acid sequence identity with mouse and rat IL-28A and shows cross-species activity. It shares 96% and 70% aa sequence identity with human IL-28B and IL-29, respectively.

  • Reconstitute at 100 μg/ml in PBS
  • Carrier-free

Type III interferons are upregulated in response to viral infection, and they act predominantly on epithelial cells through a receptor complex that contains IL-10 R beta and IL-28 R alpha /IFN-gamma R1 (4-6). Its function as anti-viral molecules and induce the upregulation of MHC class I antigen.

IL-28A promotes the Th1 polarization of dendritic cells in the airway and inhibits Th2 and Th17 mediated inflammation. It exhibits anti-tumor activity, in part by enhancing IL-12 dependent anti-tumor CTL responses in vivo. In contrast, it is up-regulated in invasive bladder cancer where it promotes tumor cell migration.

Store at –20 to –70 °C as supplied for 12 months and after reconstitution for six months; 2 to 8 °C under sterile conditions after reconstitution for one month.