Specifications
- Description:EcoMount Mounting Media
- Size:100 ml
- Cat. No.:MSPP-EM897L
- Supplier no.:EM897L
Specifications
About this item
EcoMount is a fast-drying, optically clear, permanent mounting medium. Polymer-based mounting medium does not contain hazardous reagents such as xylene, toluene or benzene as its base solvent.
- Permits longterm storage of slides with chromogens such as warp red, vulcan fast red and ferangi blue
- Can also be used with DAB and other non-soluble chromogens
- Dries quickly and retains excellent refractivity
- Compatible for use on automatic cover slipping machines
It is designed especially for the permanent preservation of horseradish peroxidase, alkaline phosphatase and Qdot (quantum dot nanocrystals) immunofluorescent techniques.