ContactUsLinkComponent
IncuCyte® Caspase-3/7 Red Apoptosis Assay Reagent, Essen Biosciences
IncuCyte® Caspase-3/7 Red Apoptosis Assay Reagent, Essen Biosciences
Catalog # MSPP-4704
Supplier:  Avantor
CAS Number:  
IncuCyte® Caspase-3/7 Red Apoptosis Assay Reagent, Essen Biosciences
Catalog # MSPP-4704
Supplier:  Avantor
CAS Number:  
Specifications
  • Description:
    IncuCyte® Caspase-3/7 Red Apoptosis Assay Reagent
  • Size:
    20 µl
  • Cat. No.:
    MSPP-4704
  • No. of wells:
    96
Specifications
About this item
IncuCyte® Caspase-3/7 red apoptosis assay reagent couples the activated caspase-3/7 recognition motif (DEVD) to NucView™488, a DNA intercalating dye to enable quantification of apoptosis over time.
  • Inert
  • Requires no wash, mix and read protocol
It is specially formulated for use in the IncuCyte® live-cell analysis system and can be added directly to tissue culture wells. It is non-perturbing to cell growth and morphology. When added to tissue culture medium, this inert, non-fluorescent substrate crosses the cell membrane where it is cleaved by activated caspase-3/7 resulting in the release of the DNA dye and red fluorescent staining of nuclear DNA. Kinetic activation of caspase-3/7 can be monitored morphologically using live cell imaging and quantified using the IncuCyte® basic analyzer.
Delivery: Supplied as 5 mm mix and read reagent in DMSO sufficient for 100 to 200 live-cell apoptosis tests in 96-well format.