Residual Solvents Class 3 - Mix A (24 Components), Restek
Catalog # 76292-048
Supplier: Restek
CAS Number:
Specifications
- Concentration:5000 ug/ml in NN-Dimethylformamide
- Description:Residual Solvents Class 3 - Mix A
- Size:1 ml/Ampule
- Description:Residual Solvents Class 3 - Mix A
- Cat. No.:76292-048
Specifications
About this item
Multicomponent mixes simplify standard prep and reduce variability. Ideal for USP 467 residual solvents analysis.
- Verified composition and stability
Certifications: Certified reference materials (CRMs) manufactured and QC tested in ISO-accredited labs satisfy your ISO requirements.