Support Adapters, Cassette Torch Injectors, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
These adapters are for ball joint or non-ball joint injectors.
Non-ball joint injectors for use with the corrosion-resistant torch and the Ryton Scott spray chamber.
Frequently Bought Together