ContactUsLinkComponent
NMR Sample Tube Holder/Rack, New Era™
NMR Sample Tube Holder/Rack, New Era™
Catalog # 100165-896
NMR Sample Tube Holder/Rack, New Era™
Catalog # 100165-896
Supplier Number:  NE3305
Specifications
  • Description:
    72-Place NMR Sample Tube Holder/Rack
  • Material:
    Polyethylene
  • Environmentally Preferable:
  • Cat. No.:
    100165-896
  • For tube O.D.:
    5 mm
  • Supplier no.:
    NE3305
Specifications
About this item
Sturdy polypropylene tube holder is ideal for staging samples during preparation, transporting samples between labs and for sample storage.
  • Rack dimensions: 8×4×9"