Specialty NMR Sample Tubes with Screw Thread, New Era™
Supplier: New ERA Enterprises
Certificates
About this item
Tubes offer the convenience of a screw cap with the security of a PTFE/silicone seal.
- Made of type 1 class B borosilicate glass
- 5 mm Ext.Ø tubes fit all standard 5 mm probes
- 10 mm Ext.Ø tubes fit all standard 10 mm probes
Each tube includes an open-top cap and a PTFE/silicone septum. A 22 gauge, standard point needle is recommended for a clean puncture. To maintain spinning quality, choose the tube length that places the threads closest to the spinner.
These tubes are ideally suited for glove box preparation of air or moisture sensitive samples. The screw cap provides easy access for kinetic studies. Another proven application has been tracking the evolution of hydrolysis products within bioabsorbable materials.
Ordering information: Optional lengths are available via special order.
Frequently Bought Together