Quartz SUPRASIL Flow-Through Cells, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
These flow-through cells are used for measuring samples with continuous flow or with sippers where individual samples are aspirated into the cell.
- All cells have center heights of 15 mm
- Semi-micro/ultra-micro cells with in- and outlet tubes
Compact with two screw connectors, M6×1 and 500 mm length FEP tubing of 1.9 mm outer diameter and 1.1 mm inner diameter.
Delivery: Each package contains one cell with two clear windows.
Frequently Bought Together