Order Entry
United States
ContactUsLinkComponent
Lindstrom® Diagonal Flush Cutter, Oval Head, JH Williams Tool Group
Lindstrom® Diagonal Flush Cutter, Oval Head, JH Williams Tool Group
Catalog # 89125-250
Lindstrom® Diagonal Flush Cutter, Oval Head, JH Williams Tool Group
Catalog # 89125-250
Supplier Number:  8141

Specifications

  • Cat. No.:
    89125-250
  • Cutting capacity:
    38 - 16 AWG
  • Overall length:
    4.33"
  • Supplier no.:
    8141
  • Cut type:
    Flush

Specifications

About this item

Lindstrom 80-Series has precision-forged and crafted oval head cutters.

  • Precision induction hardened edges 63 - 65 HR(c)
  • Carbon/chrome ball bearing steel 1%
  • Replaceable springs and precision screw joint
  • CNC machining
  • ESD safe handles

No ergonomic handle.