XYZ Goniometer Head Kits and Components, Rigaku
Supplier: Rigaku Reagents
Certificates
About this item
XYZ Goniometer Heads feature a IUCr standard goniometer mount for compatibility with most goniometers.
Each goniometer head allows ±2.5mm adjustments in X and Y and ±5mm in Z.
Delivery: Goniometer head kits are supplied with a storage case, goniometer head key, and interchangeable parts to support sample mounting with either a magnetic mount or a Z support for use with capillary or brass pin mounts.
Frequently Bought Together