CombiClover 4 Chamber 4×250 Crystallization Plates (XPT), Rigaku
Supplier: Rigaku Reagents
Certificates
About this item
CombiClover 4 Chamber Crystallization plates utilize four partitioned reservoir chambers in the place of one central reservoir. These four partitioned chambers are each connected to a satellite drop chamber via dedicated vapor diffusion channels to create a variation of the novel combinatorial 'crystallization clover' provided in the plates.
- Capacity of each of the four partitioned areas of the reservoir is 250 microliters
- Satellite drop chamber volume is 50 microliters
- Plate dimensions: 112×165×24 mm
Each plate has 24 of the partitioned reservoirs and 96 flat-bottom sitting drop chambers.
This design allows four different experiments to be conducted in each clover, with each portion containing a different crystallany, and allowing them to be investigated simultaneously.
Frequently Bought Together